Drucken
Fall protection harnesses, 1 point
Lieferant: Portwest Clothing
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
FP11RER
PRTWFP11REREA 30.2 EUR
PRTWFP11RER
Fall protection harnesses, 1 point
Fallschutz Auffanggurte
Fall Protection is put in place to prevent the risks associated with falling from heights, reducing impact force, restricting obstacle/ground collision and restricting users from fall hazard areas.
  • Dorsal D-ring
  • Chest and leg adjustment points
  • Flat bar buckles
  • Inspection cover for protection of product label
  • Retail box which aids presentation for retail sales
  • Made from polyester webbing, alloy steel, heat treated
Ideal for fall arrest or work restraint systems, our entry level full body harness is light weight and easy to use, allowing a secure fit.
Zertifizierungen: EN 361