Drucken
Fall protection harnesses, 2 point
Lieferant: Portwest Clothing
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
FP13RER
PRTWFP13REREA 49 EUR
PRTWFP13RER
Fall protection harnesses, 2 point
Fallschutz Auffanggurte
Fall Protection is put in place to prevent the risks associated with falling from heights, reducing impact force, restricting obstacle/ground collision and restricting users from fall hazard areas.
  • Superior strength
  • Fully adjustable shoulder, chest and leg straps
  • Chest loops
  • Light weight and easy to use
  • Made from polyester webbing, alloy steel, heat treated
Features front and back D-rings and is widely used in the construction industry.
Zertifizierungen: EN 361