Drucken
Recirculating chiller, SONICATOR®
Lieferant: QSONICA LLC
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
4905-220
QSON4905-220EA 7390 EUR
QSON4905-220
Recirculating chiller, SONICATOR®
Umwälzthermostate Geschlossene Umwälzsysteme Kälteumwälzthermostate
Sonication generates heat which may be detrimental to some applications. Attempting to control temperature with ice and/or repeatedly changing out water is tedious and no longer necessary. This chiller automates the sample cooling process.
  • Quick-connect tubing and fittings attach the chiller to the ports on the cup horn or microplate horn
  • When used in conjunction with the pulsed sonication mode, desired water temperature will be maintained
Older model cup horns may require special fittings.
Bestellinformation: Quick-connect tubing and fittings should be ordered separately.