Drucken

Rhodamine 123 fluoreszierender Farbstoff, Mitochondrien-Färbemittel

Lieferant: SIGMA ALDRICH MICROSCOPY
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: von 5


MQ100

R8004-100MG R8004-25MG R8004-5MG
R8004-100MGEA 635 EUR
R8004-100MG R8004-25MG R8004-5MG
Rhodamine 123 fluoreszierender Farbstoff, Mitochondrien-Färbemittel
Rhodamine 123
Formel: C₂₁H₁₇ClN₂O₃
MW: 380.82 g/mol
Storage Temperature: Room Temperature
MDL Number: MFCD00012664
CAS-Nummer: 62669-70-9
https://qa.digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8715b9096798f275fea230ef7ef997c48f67619c&vid=8715b9096798f275fea230ef7ef997c48f67619c&prid=web&clid=SAPDAM

Order Now

Spezifikation Testergebnisse

Appearance (Form) Powder
Infrared spectrum Conforms to Structure
Carbon (dry basis) 62.2 - 70.2 %
Nitrogen (dry basis) 6.9 - 7.8 %
Water (by Karl Fischer) ≤10.0 %
Solubility (Turbidity)
1mg/ml in Ethanol
Clear to Slightly Hazy
Solubility (Colour) Conforms to Requirements
Yellow-Orange
Recommended Retest Period 4 Years

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR