Drucken
Complement reagents
Lieferant: Rockland Immunochemicals
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
C302-0010 C202-0005 C220-0002 C303-0010 C303-0050 C203-0005 C304-0010 C304-0050 C304-0100 C304-0500 C204-0005 C305-0010 C305-0050 C305-0100 C309-0050 C309-0100 C209-0005 C300-0050 C300-0100 C300-0500 C200-0005 C306-0010 C306-0050 C306-0100 C206-0005 C301-0010 C301-0100 C201-0005 C205-0005 C301-0050
ROCKC302-0010EA 1100 EUR
ROCKC302-0010 ROCKC202-0005 ROCKC220-0002 ROCKC303-0010 ROCKC303-0050 ROCKC203-0005 ROCKC304-0010 ROCKC304-0050 ROCKC304-0100 ROCKC304-0500 ROCKC204-0005 ROCKC305-0010 ROCKC305-0050 ROCKC305-0100 ROCKC309-0050 ROCKC309-0100 ROCKC209-0005 ROCKC300-0050 ROCKC300-0100 ROCKC300-0500 ROCKC200-0005 ROCKC306-0010 ROCKC306-0050 ROCKC306-0100 ROCKC206-0005 ROCKC301-0010 ROCKC301-0100 ROCKC201-0005 ROCKC205-0005 ROCKC301-0050
Complement reagents
Seren
Complement reagents are prepared from healthy, non-medicated animals using very large pools to reduce lot-to-lot variability. The fresh frozen complement is rapidly processed and maintained at –70 °C to ensure maximum activity. The lyophilised complement is supplied with a diluent of veronal buffered saline containing optimal concentrations of magnesium and calcium.