Drucken
Scoops, long handled, PharmaScoop®
Lieferant: Sampling Systems
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
A396-1000
SAMPA396-1000EA 323 EUR
SAMPA396-1000
Scoops, long handled, PharmaScoop®
Schaufeln
The long handled PharmaScoop made from 316 stainless steel is ideal for use with large containers.
  • 600 mm long handle allows the operator to safely reach deep into containers.
  • Crevice-free
  • Radiused corners for easy cleaning
  • GMP correct
  • BSE/TSE-free
Body length: 235 mm
Handle length: 600 mm