Drucken
Micro tubes with thread
Lieferant: SARSTEDT
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
72.693.465 72.730.217
SARS72.693.465EA 223 EUR
SARS72.693.465 SARS72.730.217
Micro tubes with thread
Röhrchen Mikroröhrchen
PP, translucent with PP screw cap.
  • With printed label
  • Withstand centrifugation up to 20000×g
  • Attached, not screwed-on cap
  • Sterile and free of DNA, DNase/RNase, PCR inhibitors, ATP and pyrogens/endotoxins
With or without skirted base.