Drucken
Secure sharp container Multi-Safe sani 200
Lieferant: SARSTEDT
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
77.3890.002
SARS77.3890.002EA 210 EUR
SARS77.3890.002
Secure sharp container Multi-Safe sani 200
Behälter Abfallbehälter für spitze/scharfe Abfälle
Boxes, made of PP, yellow/red. Secure sharp container with final closure in a handy size for doctors, nurses, rescue services- and ambulatory medical services, or for the doctors and rescue services supply bag.
  • Single-handed opening and closing of day lid
  • U-Shaped recess for sharps removal from Luer lock systems
Zertifizierungen: TRBA 250, UN, DIN EN ISO 23907