Drucken
Sharps disposal container Multi-Safe steri
Lieferant: SARSTEDT
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
77.3899.060
SARS77.3899.060EA 1770 EUR
SARS77.3899.060
Sharps disposal container Multi-Safe steri
Behälter Abfallbehälter für spitze/scharfe Abfälle
Box, made of PP, yellow/red. Secure and robust container for the disposal of infectious wastes that can be deactivated by autoclaving at 121 or 134 °C.
  • The small cover melts at about 90°C to enable autoclaving. A grate prevents waste from falling out
  • The box is stackable both empty and filled
  • Robust handle in the lid and two moulded recesses in the box