Drucken
ThermoSafe® ChillTherm® Pre-Qualified 2 to 8 °C Shippers
Lieferant: Sonoco Thermosafe
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
CTR33
SCATCTR33EA 1120 EUR
SCATCTR33
ThermoSafe® ChillTherm® Pre-Qualified 2 to 8 °C Shippers
Versandtaschen und Transportbehälter
ChillTherm® pre-qualified 2 to 8 °C shippers are a seasonal, water-based solution which provides a cost-effective solution for the shipping of temperature-sensitive products.
  • Pre-qualified 2 to 8 °C shipper
  • Tested to several ambient profiles to speed up implementation
ChillTherm® shippers provide an economic solution when shipping products which need to be kept between 2 and 8 °C. Ideal for pharmaceuticals, biologics, medical devices, or animal health products, these shippers are easy to condition and pack out. Reports readily available so no need for lengthy testing and qualificiaton projects.