Drucken
PolarPack® Foam Brick, Ice Pack Refrigerant Ice Bricks
Lieferant: Sonoco Thermosafe
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
FPP24
SCATFPP24EA 3240 EUR
SCATFPP24
PolarPack® Foam Brick, Ice Pack Refrigerant Ice Bricks
Kühlakkus
PolarPack® gel brick coolant pack refrigerants provide thermal solutions for temperature critical packages during transit and are an alternative to wet ice packs.
  • Broad selection available
  • Reliable temperature protection
  • Never change shape during freezing
Available in a broad selection of styles and a multitude of sizes, they’re designed to provide reliable temperature protection for a vast array of products and packages.
These semi-rigid foam bricks are vacuum-sealed for consistent weight and shape. These semi-rigid foam bricks never change shape during freezing, thawing or transit, enabling superior surface contact for more consistent temperature control.