Drucken
Kimble™ ClearScrew Thread Sample Vial with Attached PTFE-Faced/White Rubber Lined Cap
Lieferant: DWK Life Sciences
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
SP60811B-1
SCERSP60811B-1EA 499 EUR
SCERSP60811B-1
Kimble™ ClearScrew Thread Sample Vial with Attached PTFE-Faced/White Rubber Lined Cap
Vials
Expansion borosilicate glass vials with attached black phenolic caps and PTFE-faced white rubber liners.
  • Small case quantities
  • Manufactured from clear 33 expansion, borosilicate glass
  • Attached closed top black phenolic closures
Zertifizierungen: USP Type I and ASTM E438, Type I, Class A requirements.