Fastpek Allergen ELISA Kits
Lieferant: HYSERVE
Ratings:
(keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
1500005
1500007-2
SCIL1500005EA
351
EUR
SCIL1500005
SCIL1500007-2
Fastpek Allergen ELISA Kits
Klinisches Diagnosezubehör
Klinische Reagenzien und Kits
These kits are designed to detect allergens by special extraction buffer and antibodies.
- High sensitivity
- Increased specificity
- Assay time of about 120 minutes
Suitable for use in highly processed food.