Silicone DC 710 fluid
Lieferant: SERVA
Ratings:
(keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
35149.04
SERA35149.04EA
482
EUR
SERA35149.04
Silicone DC 710 fluid
Flüssigkeiten für Umwälzbäder
- Retains its properties after continuous use at temperatures up to 260°C in closed systems
- Shows very low volatility even at elevated temperatures
- Highly resistant to oxidation and gumming