Drucken

Ferrozine mono-sodium salt hydrate 97%, Sigma-Aldrich®

Lieferant: SIGMAALDRICH
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: von 5

Achtung


M-Clarity™ Qualitätsstufe = MQ200

160601-1G 160601-5G 160601-25G
SIAL160601-1GEA 80.2 EUR
SIAL160601-1G SIAL160601-5G SIAL160601-25G
Ferrozine mono-sodium salt hydrate 97%, Sigma-Aldrich®
Ferrozine mono-sodium salt hydrate

Used as a colorimetric indicator for ferroxidase assay. Reagent for the determination of Fe and Co, and Ag, Cu, and Ni.

Formel: C₂₀H₁₃N₄NaO₆S₂·H₂O
MW: 492.47 g/mol (anhy)
MDL Number: MFCD00078417
CAS-Nummer: 63451-29-6

Order Now

Spezifikation Testergebnisse

Appearance (Color) Light Yellow to Very Dark Yellow and Light Green-Yellow to Very Dark Green-Yellow
Appearance (Form) Powder or Chunks
Infrared Spectrum Conforms to Structure
Carbon 47.1 - 50.5%
Water (by Karl Fischer) 3.0 - 8.0 %
Solubility (Turbidity) 50 mg/ml in H₂O Clear to Very Hazy


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR