Drucken

Rhodamin B Isothiocyanat mixed isomers, powder, BioReagent protein labeling_suitable, Sigma-Aldrich®

Lieferant: SIGMAALDRICH
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: von 5

Gefahr

R1755-100MG R1755-500MG R1755-1G
SIALR1755-100MGEA 133 EUR
SIALR1755-100MG SIALR1755-500MG SIALR1755-1G
Rhodamin B Isothiocyanat mixed isomers, powder, BioReagent protein labeling_suitable, Sigma-Aldrich®
Rhodamin B Isothiocyanat

Rhodamine B isothiocyanate (RhBITC) is an organic fluorescence dye and is widely used as a fluorescent tag in a variety of biological applications.


RhBITC is adsorbed onto Ag on silica or polystyrene beads. It exhibits not only a strong surface-enhanced Raman scattering (SERS) signal but also a measurable amount of fluorescence. The RhBITC-modified Ag-deposited silica or polystyrene beads disperse well in ethanol. They are also readily coated in water with polyelectrolytes for their further derivatization with biological molecules of interest that can bind to target molecules.


Rhodamine B isothiocyanate is used for the following applications:
• Rhodamine isothiocyanate labeling
• Rhodamine-labeled synthetic APF
• Preparations of protein and fluorescent conjugates
• Fluorescent red dye

Formel: C₂₉H₃₀ClN₃O₃S
MW: 536.08 g/mol
Storage Temperature: Gefrierschrank
MDL Number: MFCD00136007
CAS-Nummer: 36877-69-7
EINECS: 253-248-9
UN: 3335
ADR: 9 III

Order Now

Spezifikation Testergebnisse

labeling efficiency with bovine albumin > 70%


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR