Drucken

Wasser, BioReagent Cell Culture, Sterile-Filtered, Sigma-Aldrich®

Lieferant: SIGMAALDRICH
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: von 5


MQ300

W3500-100ML W3500-1L W3500-24X500ML W3500-500ML W3500-6X1L W3500-6X500ML
SIALW3500-100MLEA 17.1 EUR
SIALW3500-100ML SIALW3500-1L SIALW3500-24X500ML SIALW3500-500ML SIALW3500-6X1L SIALW3500-6X500ML
Wasser, BioReagent Cell Culture, Sterile-Filtered, Sigma-Aldrich®
Wasser

This product is a very low endotoxin (less than 1 EU/mL), 0.2 micron filtered.


For use in the preparation of cell culture media, and cell suspension and washing solutions.

Formel: H₂O
MW: 18.02 g/mol
Siedepunkt: 100
Melting Pt: 0
Dichte: 1 g/cm³ (4 °C)
Storage Temperature: Raumtemperatur
MDL Number: MFCD00011332
CAS-Nummer: 7732-18-5
EINECS: 231-791-2
Merck Index: 13,10098

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR