Drucken
Digital wood moisture analyser
Lieferant: TQC
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
LI7000
SIMELI7000EA 501 EUR
SIMELI7000
Digital wood moisture analyser
Analysatoren Feuchte-Analysatoren
Non destructive instrument for measuring moisture levels in wood.
  • Calibration lines for 25 different types of wood
  • High degree of accuracy
  • Automatic power off
The selected wood and the measuring results are immediately shown. The instrument can also detect the moisture content of concrete, anhydrite, estrich, polyester.
Lieferumfang: Supplied in a pouch.