Drucken
DNA marker, pBR322 DNA/BsuRI (HaeIII) marker
Lieferant: FERMENTAS
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
SM0271
SM0271EA 67.75 EUR
SM0271
DNA marker, pBR322 DNA/BsuRI (HaeIII) marker
Elektrophorese-Reagenzien Molekulargewichtsmarker
pBR322 DNA/BsuRI (HaeIII) marker is suitable for sizing and approximate quantification of small linear double-stranded DNA fragments in agarose and non denaturing polyacrylamide gels.
  • Agarose gel compatible
  • Sharp bands
Lieferumfang: Supplied with loading dye for sample DNA.
Achtung! Bitte beachten: For research use only. Not for use in diagnostic procedures.