DNA ladder, low range, MassRuler
Lieferant: FERMENTASGesamtbewertung: 0
Bewertung: von 5
MassRuler Low Range DNA Ladder, ready to use, 80 to 1031 bp, is specially designed for accurate quantification and sizing of DNA fragments by agarose gel electrophoresis.
- Sharp bands
- Easy to remember fragment sizes and quantities
- Premixed with 6X MassRuler DNA Loading Dye for direct loading and room temperature storage
The intensity of the fragment in each ladder is calibrated against a standard that guarantees the precise quantity of each band. The ladders are mixtures of chromatography-purified, individual DNA fragments.
Lieferumfang: Supplied with 1 ml 6X loading dye for sample DNA.