Drucken
DNA ladders, ultra low range, FastRuler
Lieferant: FERMENTAS
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
SM1103
SM1103EA 84.5 EUR
SM1103
DNA ladders, ultra low range, FastRuler
Elektrophorese-Reagenzien Molekulargewichtsmarker
FastRuler ultra low range DNA ladder is specifically designed for fast sizing and approximate quantification of double-stranded DNA in 48 well (or 96-well) high throughput gels, as well as in conventional agarose gels. The ladder is a mixture of five blunt-ended chromatography-purified individual DNA fragments that are easily resolved in a short separation distance (10 to 20 mm) after an 8 to 14 minutes run on the appropriate agarose gel.
  • Especially useful for electrophoresis of PCR products
  • Fast separation (8 to 14 min)
  • Short separation distance (10 to 20 mm)
  • Sharp bands, easy-to-remember fragment sizes and quantities
  • Ready to use – premixed with loading dye
  • Supplied with loading dye solution for sample DNA
FastRuler ultra low range DNA ladder contains dephosphorylated DNA fragments and is ideal for forward 5-end labeling with T4 polynucleotide kinase.