Drucken

Choline dihydrogen citrate ≥98%

Lieferant: SPECTRUM CHEMICAL MFG
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: von 5

C1229-12KG C1229-25KGBL C1229-50KGBL
SPCMC1229-12KGEA 3670 EUR
SPCMC1229-12KG SPCMC1229-25KGBL SPCMC1229-50KGBL
Choline dihydrogen citrate ≥98%
Choline dihydrogen citrate

Choline Dihydrogen Citrate is part of a class of quaternary ammonium salts containing the N,N,N-trimethylethanolammonium cation and Dihydrogen Citrate counteranion. Choline is the precursor for the neurotransmitter acetylcholine, which is involved in many functions including memory and muscle control. This item is of known assay and suitable for most chemical uses.


GHS hazard : Exclamation mark

Formel: C₁₁H₂₁NO₈
MW: 295.29 g/mol
Storage Temperature: Raumtemperatur
MDL Number: MFCD00038734
CAS-Nummer: 77-91-8

Order Now

Spezifikation Testergebnisse

Assay 98% min.
Water max.0.25 %
Residue On Ignition max. 0.05 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR