Drucken

trans-1,2-Diaminocyclohexan-N,N,N',N'-tetraessigsäure Monohydrat ≥98%, Reagenzqualität

Lieferant: SPECTRUM CHEMICAL MFG
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: von 5

Achtung

C1466-12KGBL
SPCMC1466-12KGBLEA 0 EUR
SPCMC1466-12KGBL
trans-1,2-Diaminocyclohexan-N,N,N',N'-tetraessigsäure Monohydrat ≥98%, Reagenzqualität
trans-1,2-Diaminocyclohexan-N,N,N',N'-tetraessigsäure Monohydrat

(1,2-Cyclohexylenedinitrilo)tetraacetic Acid, Reagent is an alkyl-substituted amino acid that functions as a chelating agent. This ingredient's source is synthetic. It appears as a solid and is partially soluble in cold water. Its reagent grade means this is the highest quality commercially available for this chemical and that the American Chemical Society has not officially set any specifications for this material.

Formel: C₁₄H₂₂N₂O₈·1H₂O
MW: 364.35 g/mol
Melting Pt: 213…216 °C
Storage Temperature: Raumtemperatur
MDL Number: MFCD00066429
CAS-Nummer: 125572-95-4
EINECS: 236-308-9

Order Now

Spezifikation Testergebnisse

ASSAY (C14H22N2O8•H2O) 98% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR