Drucken

Folic acid ≥97,0%, Pulver USP

Lieferant: SPECTRUM CHEMICAL MFG
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: von 5

FO105-1KG FO105-25KGBL FO105-5KGBL FO105-100GM
SPCMFO105-1KGEA 4400 EUR
SPCMFO105-1KG SPCMFO105-25KGBL SPCMFO105-5KGBL SPCMFO105-100GM
Folic acid ≥97,0%, Pulver USP
Folsäure

All Spectrum Chemical USP products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formel: C₁₉H₁₉N₇O₆
MW: 441.41 g/mol
Storage Temperature: Raumtemperatur
MDL Number: MFCD00079305
CAS-Nummer: 59-30-3

Order Now

Spezifikation Testergebnisse

Assay (Anhydrous Basis) 97.0 - 102.0%
Water Determination max.8.5 %
Residue On Ignition max. 0.3 %
Related Compounds max. 2.0 %
Identification To Pass Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR