Drucken

HEPES (2-[4-(2-Hydroxyethyl)-1-piperazinyl]ethansulfonsäure) ≥99,5% (durch titrimetrische Analyse), cGMP Herstellung

Lieferant: SPECTRUM CHEMICAL MFG
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: von 5

H1804-12KGBL H1804-1KGBL H1804-25KGBL
SPCMH1804-12KGBLEA 16760 EUR
SPCMH1804-12KGBL SPCMH1804-1KGBL SPCMH1804-25KGBL
HEPES (2-[4-(2-Hydroxyethyl)-1-piperazinyl]ethansulfonsäure) ≥99,5% (durch titrimetrische Analyse), cGMP Herstellung
HEPES (2-[4-(2-Hydroxyethyl)-1-piperazinyl]ethansulfonsäure)

HEPES, cGMP Manufactured is a zwitterionic biological buffer and is one of Good's buffers widely used in cell culture. Non-graded HEPES, cGMP Manufactured supplied by Spectrum is indicative of a grade suitable for general industrial use or research purposes and typically are not suitable for human consumption or therapeutic use.

Formel: C₈H₁₈N₂O₄S
MW: 238.31 g/mol
Storage Temperature: Raumtemperatur
MDL Number: MFCD00006158
CAS-Nummer: 7365-45-9
EINECS: 230-907-9
Merck Index: 13,04672

Order Now

Spezifikation Testergebnisse

APPEARANCE WHITE FINE POWDER
ASSAY (TITRATION) min. 99.5 %
pH OF A 5% AQUEOUS SOLUTION 4.5 - 6.5
pKa @ 25°C 7.35- 7.75
Absorbance @ 260 nm (0.05 M in water) max. 0.05 AU
Absorbance @ 280 nm (0.05 M in water) max. 0.03 AU
WATER (KF) max. 0.5 %
HEAVY METALS (as Pb) max. 5 ppm
INSOLUBLE MATTER max. 0.1 %
TOTAL AEROBIC MICROBIAL COUNT max. 100 cfu/g
TOTAL YEAST AND MOLD COUNT max. 100 cfu/g
BACTERIAL ENDOTOXIN As reported


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR