2-Ethylhexyl salicylate ≥95% USP
Lieferant: SPECTRUM CHEMICAL MFG
Ratings:
(keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
O1236-100ML
SPCMO1236-100MLEA
168
EUR
SPCMO1236-100ML
2-Ethylhexyl salicylate ≥95% USP
2-Ethylhexylsalicylat
Octisalate, USP absorbs ultraviolet radiation in the UVB range and is used in sunscreens and other cosmetics to protect from skin damage. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Formel:
C₁₅H₂₂O₃ MW: 250.34 g/mol Storage Temperature: Raumtemperatur |
MDL Number:
MFCD00053300 CAS-Nummer: 118-60-5 EINECS: 204-263-4 |
Spezifikation Testergebnisse
Assay | 95% Min. |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...