Drucken

Potassium iodide ≥99,0%, körnig FCC

Lieferant: SPECTRUM CHEMICAL MFG
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: von 5

P1344-2.5KG P1344-500GM
SPCMP1344-2.5KGEA 3250 EUR
SPCMP1344-2.5KG SPCMP1344-500GM
Potassium iodide ≥99,0%, körnig FCC
Kaliumiodid

Potassium Iodide, Granular, FCC is used as a source for dietary iodine. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.


GHS hazard : Exclamation mark;Health hazard

Formel: Kl
MW: 166 g/mol
Siedepunkt: 1330
Melting Pt: 686
Storage Temperature: Raumtemperatur
MDL Number: MFCD00011405
CAS-Nummer: 7681-11-0
Merck Index: 13,07727

Order Now

Spezifikation Testergebnisse

Assay (Dried Basis) 99.0 - 101.5%
Iodate (IO₃) max. 4 mg/kg
Lead (Pb) max. 4 mg/kg
Loss On Drying max. 1 %
Nitrate, Nitrite, and Ammonia To Pass Test


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR