Drucken

Polyethylene glycol 4500 NF

Lieferant: SPECTRUM CHEMICAL MFG
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: von 5

P1792-12KG P1792-2.5KG
SPCMP1792-12KGEA 1060 EUR
SPCMP1792-12KG SPCMP1792-2.5KG
Polyethylene glycol 4500 NF
Polyethylenglycol 4.000

Polyethylene Glycol 4500, NF acts as a lubricant, coating the surfaces in aqueous and non-aqueous environments.


All Spectrum Chemical NF grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP).

Formel: H(OCH₂CH₂)nOH
MW: 4500 g/mol
Storage Temperature: Raumtemperatur
MDL Number: MFCD00081839
CAS-Nummer: 25322-68-3

Order Now

Spezifikation Testergebnisse

Molecular Weight 4050-4950
Completeness and Color of Solution To Pass Test
Viscosity @98.9 C (210 F) +/-0.3 C 140- 200 Cs
pH of a 5% Solution @ 25°C 4.5- 7.5
Residue On Ignition max. 0.1 %


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR