Drucken

Sodium bisulphite, körnig FCC

Lieferant: SPECTRUM CHEMICAL MFG
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: von 5

Gefahr

S1172-2.5KG S1172-500GM
SPCMS1172-2.5KGEA 389 EUR
SPCMS1172-2.5KG SPCMS1172-500GM
Sodium bisulphite, körnig FCC
Natriumhydrogensulfit

Sodium Bisulfite, Granular, FCC is used as an antioxidant to preserve the aromas and flavors. The FCC grade meets the requirements of the Food Chemical Codex indicates and is suitable for all food, beverage and nutritional supplement applications. Spectrum Chemical offers over 300 Food grade chemical ingredients packaged in laboratory size bottles to production drum quantities and are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Formel: HNaO₃S
MW: 104.06 g/mol
Melting Pt: 300 °C
Dichte: 2,476 g/cm³ (20 °C)
Storage Temperature: Raumtemperatur
MDL Number: MFCD00167602
CAS-Nummer: 7631-90-5
EINECS: 231-548-0
UN: 2693
ADR: 8 III
Merck Index: 12,08731

Order Now

Spezifikation Testergebnisse

Assay 58.5% Min.


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR