Drucken
Microfiltration filters, MiniKap®
Lieferant: Spectrum Laboratories
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
MK2M204V6N
SPECMK2M204V6NEA 847 EUR
SPECMK2M204V6N
Microfiltration filters, MiniKap®
Filter Membranfilterkapseln
These Point-of-use microfiltration filters are excellent for water and gas filtration systems. MiniKap® HF filter provides the filtration efficiency of the 0,2 µm DynaFibre® and air vent fibre in a device conveniently equipped with a ¼'' MNPT, Luer-Lok or hose barb inlet connection.