Drucken
DNA ladders and markers
Lieferant: STRATAGENE
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
201115
STRA201115EA 383 EUR
STRA201115
DNA ladders and markers
Elektrophorese-Reagenzien Molekulargewichtsmarker
The supercoiled DNA ladder contains fragments from 2 to 10 kB at 1 kB increments and an additional band at 12 kB.
  • Fragments are exact multiples or fractions of 1000 bp
  • Includes useful reference point for determining location
  • Economical
  • Easy radiolabelling of ladder
  • Known mass amount of each fragment for DNA quantitation
The kB DNA ladder contains fragments from 1 to 10 kB at 1 kB increments, as well as bands at 250 bp, 500 bp, 750 bp, 1500 bp and 12 kB.
Concentration: 500 µg/ml in 10 mM Tris-HCl (pH 7,5), 1 mM EDTA.
Achtung! Bitte beachten: For research use only. Not for use in diagnostic procedures.
Order Now
TECHNISCHE DATEN
Environmentally Preferable