Drucken
Cis-reporting systems, PathDetect
Lieferant: STRATAGENE
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
219059 219071 219074 219080 219082 219083 219084 219085 219087 219089 219090 219091 240039
STRA219059EA 669 EUR
STRA219059 STRA219071 STRA219074 STRA219080 STRA219082 STRA219083 STRA219084 STRA219085 STRA219087 STRA219089 STRA219090 STRA219091 STRA240039
Cis-reporting systems, PathDetect
Mikroarray-Reagenzien
The PathDetect cis-reporting systems is designed for rapid and convenient assessment of the in vivo activation of these pathways. Each cis-reporter plasmid contains the luciferase reporter gene driven by a basic promoter element (TATA box) joined to tandem repeats of various binding elements.
  • Identify pathway-specific transcription activation
  • Easier and faster than blotting or gel-shift assays
  • Sensitive, quantitative assays
When PathDetect cis-reporting and expression plasmids encoding the gene of interest are co-transfected into mammalian cells, increased reporter activity indicates transcription activation and involvement of your gene product in the pathway leading to the enhancer element.
Achtung! Bitte beachten: For research use only. Not for use in diagnostic procedures.