Drucken
Stitch cutters
Lieferant: SWANN MORTON
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
0420 0421
SWAN0420EA 52.5 EUR
SWAN0420 SWAN0421
Stitch cutters
Skalpelle Skalpelle, komplett
These sterile stitch cutters are made from either carbon steel or stainless steel.
  • Convenient and cost effective
  • For use without a handle
The traditional carbon steel standard stitch cutter with its rounded bull nose tip which ensures there is no damage to the skin whilst the surgical edge cuts through the suture with minimal resistance.
The long stitch cutter allows for a more aseptic techinique by operating further away from the actual wound which also permits better visibility