Drucken
Handle for craft tool
Lieferant: SWANN MORTON
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
1235
SWAN1235EA 39 EUR
SWAN1235
Handle for craft tool
Skalpelle Skalpellgriffe
Handle made from impact resistant PS.
  • A thumbscrew is used for blade changing and retaining
  • For use with blades No. 1, 2 or 3
This craft tool is an ideal tool for marquetry, modelling, craft, studio work and horticulture.
Bestellinformation: Handle only. Blades need to be ordered separately.