Drucken
MagicPrep™ NGS System
Lieferant: TECAN TRADING
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
30186621 30186622 30186623 30186624 30186625 30186626 30186627 30186628 30186629 30191459 30191458 30191460 30191465
TCAT30186621EA 0 EUR
TCAT30186621 TCAT30186622 TCAT30186623 TCAT30186624 TCAT30186625 TCAT30186626 TCAT30186627 TCAT30186628 TCAT30186629 TCAT30191459 TCAT30191458 TCAT30191460 TCAT30191465
MagicPrep™ NGS System
Next Generation Sequencing Instruments
MagicPrep™ NGS system is a new category of automated laboratory equipment that provides push button simplicity for NGS library preparation. The Revelo DNA-Seq Enz for MagicPrep™ NGS library preparation kit provides all the reagents for generating DNA-Seq libraries on the MagicPrep™ NGS system.
  • Simple run setup in 10 minutes
  • Reliable and reproducible
  • True walk-away automation
  • Pre-optimised DNA fragmentation for a broad input range simplifies the workflow
  • Provides even coverage and uniformity across the bacterial genome regardless of GC content
  • High quality libraries from pre-fragmented DNA
  • Fully automated workflow
  • Start with total RNA and MagicPrep™ NGS does all the library preparation steps, including selection of polyadenylated transcripts
  • Even coverage across the entire transcript enabling detection of splice variants and isoforms during data analysis
  • High quality data with consistent gene detection across a 100-fold difference in input
MagicPrep™ NGS system is a new category of automated laboratory equipment that provides push button simplicity for NGS library preparation for Illumina sequencing platforms.The MagicPrep™ NGS system provides a complete solution that includes the instrument, software, pre-optimized scripts, reagents and consumables.The MagicPrep™ NGS system integrates everything you need for library preparation; all you need to do is add your samples.
The Revelo DNA-Seq Enz for MagicPrep™ NGS library preparation kit (sold separately) provides all the reagents for generating DNA-Seq libraries on the MagicPrep™ NGS system. This kit features a broad input range from 50 to 500 ng of intact genomic DNA, robust enzymatic DNA fragmentation and UDI adaptors for flexible multiplexing. This kit together with the MagicPrep™ NGS system provides a fully automated solution for DNA-Seq library preparation with a 10 minute setup time.
The Revelo mRNA-Seq for MagicPrep™ NGS library preparation kit (sold separately) provides all the reagents for generating stranded mRNA-Seq libraries on the MagicPrep™ NGS system. This kit features a broad input range from 10 to 1000 ng of total RNA and UDI adaptors for flexible multiplexing. This kit together with the MagicPrep™ NGS system provides a fully automated solution for mRNA-Seq library preparation with a 10 minute setup time.