Drucken
Microplate washer, HydroFlex™ Plus
Lieferant: TECAN TRADING
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
91100001
TCAT91100001EA 0 EUR
TCAT91100001
Microplate washer, HydroFlex™ Plus
Mikrotiterplatten-Washer
The HydroFlex Plus microplate washer is a truly flexible and upgradeable platform for automated microplate strip washing n 96-well plate formats. It is an ideal solution for a wide range of cell- and ELISA-based applications.
  • Control of wash head positions and speed settings for gentle cell washing
  • Simultaneous aspiration and dispensing for ELISA washing
  • Up to two aspiration points per well for minimal residual volumes
  • Ready for robotic integration
  • Automated buffer switching of four liquids and plate shaking capability
  • Predefined plate library and