Drucken
Cole-Parmer® WBU-200 Series Liquid Calibration Baths
Lieferant: Cole-Parmer
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
16104-50 16105-04 16112-03
TECN16104-50EA 1863.15 EUR
TECN16104-50 TECN16105-04 TECN16112-03
Cole-Parmer® WBU-200 Series Liquid Calibration Baths
Umwälzthermostate Offene Umwälzsysteme Kalibrier-Thermostate
Compact and accurate water baths for external circulation or temperature calibration.
  • Fully insulated baths offer excellent heat retention
  • Temperature stability of ±0.005 °C depending on choice of control unit
These compact calibration baths offer excellent stability over the entire temperature range. Use these
baths for external circulation to maintain temperatures of samples in viscometers, photometers, refractometers, fermenters, and other reaction vessels.
All models offer high pump performance and exceptional thermal stability from −35 to 250 °More Product Information