Drucken

Savant™ SpeedVac™ SPD2030P1 Integrated Vacuum Concentrator System Kit

Lieferant: THERMO SAVANT
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: von 5

SPD2030P1-230
THSASPD2030P1-230EA 28400 EUR
THSASPD2030P1-230
Savant™ SpeedVac™ SPD2030P1 Integrated Vacuum Concentrator System Kit
Konzentratoren
Dry and concentrate aqueous or non-aggressive samples with speed and efficiency with Thermo Scientific™ Savant SPD1030 and SPD2030 integrated SpeedVac™ systems and kits.

  • Proven reliability: Over 50 years of experience with an oil-free PTFE diaphragm vacuum pump for virtually maintenance-free operation
  • Speed and efficiency: Rapid evapouration and solvent removal with a radiant cover and vacuum controls to prevent sample bumping
  • Ease of use: Intuitive design with a bright display, three preset and three user-defined programs, and an oil-free vacuum source. USB port for data transfer
  • Safety assurance: Soft-close glass lids lock securely for the safety of samples and technicians

Ideal for drying DNA/RNA precipitates, PCR samples, proteins, enzyme, HPLC fractions in plates or tubes in water/ethanol or isopropanol and other organic solvents. SpeedVac SPD1030 system is designed for medium capacity (i.e. 120×1,5 ml tubes, other formats). SpeedVac SPD2030 system allows higher capacity (i.e. 200×1,5 ml tubes, other formats).

Solvent and solvent combinations: Acetonitrile, ethanol, methanol, methylene chloride, water, other organic solvents

Bestellinformation: SPD2030P1 Kit includes: Integrated SpeedVac system, 200×1,5 ml rotor (RH200-12), CryoCool™ 1 L heat transfer fluid, glass condensation flask (GCF400).
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR