Drucken

Anti-LGALS7 Rabbit Polyclonal Antibody (PE (Phycoerythrin))

Lieferant: US Biological
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: von 5

037748-PE
USBI037748-PEEA 884 EUR
USBI037748-PE
Anti-LGALS7 Rabbit Polyclonal Antibody (PE (Phycoerythrin))
Antikörper
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-500
Do not freeze!

Type: Primary
Antigen: LGALS7 (lectin, galactoside-binding, soluble, 7)
Clonality: Polyclonal
Clone:
Conjugation: PE (Phycoerythrin)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human
Order Now
[{"stiboOrderingTable":"<!--Specifications Tab-->\r\n\t\t\t\t<h2>TECHNISCHE DATEN</h2>\r\n\t\t\t\t <div id="stiboSpecGrid">\r\n\t\t\t\t\t<table id="stiboSpecGridTable" width="100%" class="productGrid">\r\n\t\t\t\t\t\t<caption class="caption">\r\n\t\t\t\t\t\t</caption>\r\n\t\t\t\t\t\t<tbody>\r\n\t\t\t\t\t\t\t<tr>\r\n\t\t\t\t\t\t\t\t<th></th>\t\t\t\t\t\t\t\t\t\t\t\r\n\t\t\t\t\t\t\t</tr>\r\n\t\t\t\t\t\t\t<tr class="productGrid">\t\r\n\t\t\t<td class="productGrid bld" colspan="1">Antigen-Symbol</td>\r\n\t\t\t<td class="productGrid bld" colspan="1">LGALS7</td>\r\n\t\t\t</tr>\r\n\t\t<tr class="productGrid">\t\r\n\t\t\t<td class="productGrid bld" colspan="1">Environmentally Preferable</td>\r\n\t\t\t<td class="productGrid bld" colspan="1"></td>\r\n\t\t\t</tr>\r\n\t\t<tr class="productGrid">\t\r\n\t\t\t<td class="productGrid bld" colspan="1">Antigen-Name</td>\r\n\t\t\t<td class="productGrid bld" colspan="1">lectin, galactoside-binding, soluble, 7</td>\r\n\t\t\t</tr>\r\n\t\t<tr class="productGrid">\t\r\n\t\t\t<td class="productGrid bld" colspan="1">Antikörper-Typ</td>\r\n\t\t\t<td class="productGrid bld" colspan="1">Primär</td>\r\n\t\t\t</tr>\r\n\t\t<tr class="productGrid">\t\r\n\t\t\t<td class="productGrid bld" colspan="1">Klonalität</td>\r\n\t\t\t<td class="productGrid bld" colspan="1">Polyklonal</td>\r\n\t\t\t</tr>\r\n\t\t<tr class="productGrid">\t\r\n\t\t\t<td class="productGrid bld" colspan="1">Konjugation</td>\r\n\t\t\t<td class="productGrid bld" colspan="1">PE (Phycoerythrin)</td>\r\n\t\t\t</tr>\r\n\t\t<tr class="productGrid">\t\r\n\t\t\t<td class="productGrid bld" colspan="1">Reaktivität</td>\r\n\t\t\t<td class="productGrid bld" colspan="1">Human</td>\r\n\t\t\t</tr>\r\n\t\t<tr class="productGrid">\t\r\n\t\t\t<td class="productGrid bld" colspan="1">Wirt</td>\r\n\t\t\t<td class="productGrid bld" colspan="1">Rabbit</td>\r\n\t\t\t</tr>\r\n\t\t<tr class="productGrid">\t\r\n\t\t\t<td class="productGrid bld" colspan="1">Isotyp</td>\r\n\t\t\t<td class="productGrid bld" colspan="1">IgG</td>\r\n\t\t\t</tr>\r\n\t\t<tr class="productGrid">\t\r\n\t\t\t<td class="productGrid bld" colspan="1">Western blot</td>\r\n\t\t\t<td class="productGrid bld" colspan="1">Yes</td>\r\n\t\t\t</tr>\r\n\t\t<tr class="productGrid">\t\r\n\t\t\t<td class="productGrid bld" colspan="1">ELISA</td>\r\n\t\t\t<td class="productGrid bld" colspan="1">Yes</td>\r\n\t\t\t</tr>\r\n\t\t<tr class="productGrid">\t\r\n\t\t\t<td class="productGrid bld" colspan="1">Epitop</td>\r\n\t\t\t<td class="productGrid bld" colspan="1">internal</td>\r\n\t\t\t</tr>\r\n\t\t<tr class="productGrid">\t\r\n\t\t\t<td class="productGrid bld" colspan="1">Aminosäurezahl</td>\r\n\t\t\t<td class="productGrid bld" colspan="1">54 to 82</td>\r\n\t\t\t</tr>\r\n\t\t<tr class="productGrid">\t\r\n\t\t\t<td class="productGrid bld" colspan="1">Form</td>\r\n\t\t\t<td class="productGrid bld" colspan="1">liquid</td>\r\n\t\t\t</tr>\r\n\t\t<tr class="productGrid">\t\r\n\t\t\t<td class="productGrid bld" colspan="1">Reinigung</td>\r\n\t\t\t<td class="productGrid bld" colspan="1">purified by Protein A chromatography</td>\r\n\t\t\t</tr>\r\n\t\t<tr class="productGrid">\t\r\n\t\t\t<td class="productGrid bld" colspan="1">Lagerungspuffer</td>\r\n\t\t\t<td class="productGrid bld" colspan="1">PBS, pH 7,2</td>\r\n\t\t\t</tr>\r\n\t\t<tr class="productGrid">\t\r\n\t\t\t<td class="productGrid bld" colspan="1">Lagertemperatur</td>\r\n\t\t\t<td class="productGrid bld" colspan="1">Cool</td>\r\n\t\t\t</tr>\r\n\t\t<tr class="productGrid">\t\r\n\t\t\t<td class="productGrid bld" colspan="1">Transporttemperatur</td>\r\n\t\t\t<td class="productGrid bld" colspan="1">Ice</td>\r\n\t\t\t</tr>\r\n\t\t<tr class="productGrid">\t\r\n\t\t\t<td class="productGrid bld" colspan="1">Validierte Anwendungen</td>\r\n\t\t\t<td class="productGrid bld" colspan="1">Fluoresence ELISA</td>\r\n\t\t\t</tr>\r\n\t\t<style type="text/css">\r\n \t.stibospecprodgrid {\r\n \t\t\tdisplay:block;\r\n \t\t\toverflow-x:auto;\r\n \t\t\t}\t\r\n</style>\r\n\r\n<script type="text/javascript">\r\n\r\n\t$(document).ready(function() {\r\n\t\tif (numCol("#stiboSpecGridTable") > 4){\r\n\t\t\t$('#stiboSpecGrid').addClass('stibospecprodgrid');\r\n\t\t}\t\t\r\n\t});\r\n\t\r\n\tfunction numCol(table) {\r\n\t var maxColNum = 0;\r\n\t var i=0;\r\n\t var trs = $(table).find("tr");\r\n\t for ( i=0; i<trs.length; i++ ) {\r\n\t maxColNum = Math.max(maxColNum, getColForTr(trs[i]));\r\n\t }\r\n\r\n\t return maxColNum;\r\n\t}\r\n\tfunction getColForTr(tr) {\r\n\r\n\t var tds = $(tr).find("td");\r\n\r\n\t var numCols = 0;\r\n\r\n\t var i=1;\r\n\t for ( i=1; i<tds.length; i++ ) {\r\n\t var span = $(tds[i]).attr("colspan");\r\n\r\n\t if ( span )\r\n\t numCols += parseInt(span);\r\n\t else {\r\n\t numCols++;\r\n\t }\r\n\t }\r\n\t return numCols;\r\n\t}\r\n</script>\t\t\t\t\t\t\t\t\r\n</tbody>\r\n\t\t\t\t\t</table>\r\n\t\t\t\t\t</div>"}]

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR