Drucken

Anti-ITGB2 Rat Monoclonal Antibody (Biotin) [clone: M18/2]

Lieferant: US Biological
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: von 5

C2269-02H4
USBIC2269-02H4EA 390 EUR
USBIC2269-02H4
Anti-ITGB2 Rat Monoclonal Antibody (Biotin) [clone: M18/2]
Antikörper
Recommended dilution: Flow Cytometry:1:5;

Type: Primary
Antigen: ITGB2 (integrin, beta 2 (complement component 3 receptor 3 and 4 subunit))
Clonality: Monoclonal
Clone: M18/2
Conjugation: Biotin
Epitope:
Host: Rat
Isotype: IgG2a
Reactivity: Mouse
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR