Drucken

Anti-UHRF1BP1L Chicken Polyclonal Antibody

Lieferant: US Biological
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: von 5

K1747-70
USBIK1747-70EA 474 EUR
USBIK1747-70
Anti-UHRF1BP1L Chicken Polyclonal Antibody
Antikörper
Recommended dilution: Western Blot: 1:1000-1:2000

Type: Primary
Antigen: UHRF1BP1L (UHRF1 binding protein 1-like)
Clonality: polyclonal
Clone:
Conjugation: unconjugated
Epitope:
Host: Chicken
Isotype: IgY
Reactivity: Human
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR