Drucken

Anti-Maltose Binding Protein Mouse Monoclonal Antibody [clone: 7G34]

Lieferant: US Biological
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: von 5

M2155-09B-100
USBIM2155-09B-100EA 511 EUR
USBIM2155-09B-100
Anti-Maltose Binding Protein Mouse Monoclonal Antibody [clone: 7G34]
Antikörper
Recommended dilution: Western Blot: 1:1000-1:3000

Type: Primary
Antigen: Maltose Binding Protein
Clonality: Monoclonal
Clone: 7G34
Conjugation: unconjugated
Epitope:
Host: Mouse
Isotype: IgG1 kappa
Reactivity: Human,
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR