Drucken
Anti-UHRF1 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Lieferant: US Biological
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
U1008-01-APC
USBIU1008-01-APCEA 884 EUR
USBIU1008-01-APC
Anti-UHRF1 Rabbit Polyclonal Antibody (APC (Allophycocyanin))
Antikörper
Recommended dilution: FLISA: 1:1000; Western Blot: 1:100-1:500
Do not freeze!
Type: Primary
Antigen: UHRF1 (ubiquitin-like with PHD and ring finger domains 1)
Clonality: polyclonal
Clone:
Conjugation: APC (Allophycocyanin)
Epitope: internal
Host: Rabbit
Isotype: IgG
Reactivity: Human