Drucken
BioCleanPlate Wako™ Microplates
Lieferant: Wako
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
293-35221
WAKO293-35221EA 0 EUR
WAKO293-35221
BioCleanPlate Wako™ Microplates
Mikrotiterplatten Deep-Well-Mikroplatten
The BioCleanPlate Wako™ are endotoxin-free microplates that are individually wrapped to reduce the chance of contamination and are perfect for LAL testing.
  • Endotoxin-free 96-well microplate (<0,01 EU/plate)
  • Individually packaged, ideal for use in clean rooms
  • Packaging film features dust-free protection
  • Plates are designed for use with microplate readers
When used in conjunction with BioCleanTip Wako® Extend S allows for aliquoting multi-test reagent into Microplates accurately and precisely avoiding error or bubbles.