Drucken

Reagenzglasgestelle

Lieferant: WDF - Funke GmbH & Co. KG
Ratings: (keine Bewertung)

Gesamtbewertung: 0
Bewertung: von 5

5270-312-13 5270-412-13 527-412-11 5273-26-11 527-1010-11 5270-1010-11 527-412-13 527-212-11 527-212-13 527-212-15 5278-412-13 527-34-13 527-26-11 527-46-11
212-6433EA 180 EUR
212-6433 212-6434 WCDF527-412-11 WCDF5273-26-11 WCDF527-1010-11 WCDF5270-1010-11 WCDF527-412-13 WCDF527-212-11 WCDF527-212-13 WCDF527-212-15 WCDF5278-412-13 WCDF527-34-13 WCDF527-26-11 WCDF527-46-11
Reagenzglasgestelle
Gestelle Reagenzglasgestelle
Test tube racks in stainless steel, white PE, stainless steel or white PA.

  • PE white is heat resistant up to 60 °C
  • Stainless steel is electrolytically polished
  • PA white is permanently heat resistant up to 80 °C and briefly heat resistant up to 160 °C when surface is not mechanically stressed

These racks are 70 mm high.
Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR