Drucken
Bag closure with hanger, WeLoc® PA 70
Lieferant: WELOC
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: von 5
1507001
WELO1507001EA 50.6 EUR
WELO1507001
Bag closure with hanger, WeLoc® PA 70
Klemmen Klemmen für den allgemeinen Gebrauch
This sealing clip with with integrated ring attachment is made from high quality Polyamide 6.6 material from DuPont®. The clip helps to maintain hygiene standards, securing opened bags of raw ingredients and materials, such as powders and fluids.
  • Re-closing of partly used bags of ingredients will reduce the risk of cross-contamination
  • Reduce the risk of leakage or accidents
  • Made from strong and durable material
  • Temperature resistant from −20 to +140 °C
  • With ring for additional security
  • Manufactured in Sweden under strict hygiene conditions
  • Clips can be sterilised via autoclave, EtO or gamma irradiation before being re-used
Ideal for applications where the function of a ring attachment provides an additional security options, such as for keys or tools in the factory, home or garden workshop.
Zertifizierungen: Meets current European and US food contact legislation, with production certified to ISO/TS 16949:2009 and 14001 standards.