Drucken
Heating Mantles with stirrer WHM, for beakers 5000 ml, w. integrated power controller, temperature range 450C, 800 W, power supply 230 V 1 * 1 ST
Lieferant: WITEG LABORTECHNIK
Heating Mantles with stirrer WHM, for beakers 5000 ml, w. integrated power controller, temperature range 450C, 800 W, power supply 230 V 1 * 1 ST