Drucken
Capillary columns, TraceGOLD™ TG-225MS
Lieferant: Thermo Fisher Scientific
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
26083-1540 26083-1420 26083-1430
555-8975EA 1330 EUR
555-8975 555-8978 555-8977
Capillary columns, TraceGOLD™ TG-225MS
Chromatographiesäulen
This low-bleed, stable column produces consistent results, and the low baseline noise enables improved limits of detection with enhanced resolution of low level analytes.
  • Polar phase, 50% cyano propylmethyl/50% phenylmethyl polysiloxane
  • Innovative deactivation process for siloxane to reduce tailing and improves efficiency over comparable columns
  • Equivalent to USP G7, G19 phases
Reducing column bleed increases reproducibility of the stationary phase over time, resulting in greater run-to-run and batch-to-batch reproducibility.