Drucken
Industrial gloves, Nitrile Light Knitwrist, A330
Lieferant: Portwest Clothing
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
A330YERL A330YERM A330YERS A330YERXL A330YERXXL
PRTWA330YERLEA 25.3 EUR
PRTWA330YERL PRTWA330YERM PRTWA330YERS PRTWA330YERXL PRTWA330YERXXL
Industrial gloves, Nitrile Light Knitwrist, A330
Handschuhe
These nitrile gloves with cotton interlock lining are perfect for small parts handling.
  • Lightweight, abrasion resistant
  • ³/₄ dipped for increased protection
  • Open back (without coating) for ventilation
  • Construction of the glove helps reduce hand fatigue
Excellent for jobs requiring high dexterity.
EN 388 (4111)
Zertifizierungen: EN 388:2016 (3111X).
Order Now
[{"stiboOrderingTable":"<!--Specifications Tab-->\r\n\t\t\t\t<h2>TECHNISCHE DATEN</h2>\r\n\t\t\t\t <div id="stiboSpecGrid">\r\n\t\t\t\t\t<table id="stiboSpecGridTable" width="100%" class="productGrid">\r\n\t\t\t\t\t\t<caption class="caption">\r\n\t\t\t\t\t\t</caption>\r\n\t\t\t\t\t\t<tbody>\r\n\t\t\t\t\t\t\t<tr>\r\n\t\t\t\t\t\t\t\t<th></th>\t\t\t\t\t\t\t\t\t\t\t\r\n\t\t\t\t\t\t\t</tr>\r\n\t\t\t\t\t\t\t<tr class="productGrid">\t\r\n\t\t\t<td class="productGrid bld" colspan="1">Modell</td>\r\n\t\t\t<td class="productGrid bld" colspan="1">Nitrile Light Knitwrist A330</td>\r\n\t\t\t</tr>\r\n\t\t<tr class="productGrid">\t\r\n\t\t\t<td class="productGrid bld" colspan="1">Beidseitig tragbar / handspezifisch</td>\r\n\t\t\t<td class="productGrid bld" colspan="1">Handspezifisch</td>\r\n\t\t\t</tr>\r\n\t\t<tr class="productGrid">\t\r\n\t\t\t<td class="productGrid bld" colspan="1">Environmentally Preferable</td>\r\n\t\t\t<td class="productGrid bld" colspan="1"></td>\r\n\t\t\t</tr>\r\n\t\t<tr class="productGrid">\t\r\n\t\t\t<td class="productGrid bld" colspan="1">Einweg / wiederverwendbar</td>\r\n\t\t\t<td class="productGrid bld" colspan="1">Wiederverwendbar</td>\r\n\t\t\t</tr>\r\n\t\t<tr class="productGrid">\t\r\n\t\t\t<td class="productGrid bld" colspan="1">Trägermaterial</td>\r\n\t\t\t<td class="productGrid bld" colspan="1">Baumwolle</td>\r\n\t\t\t</tr>\r\n\t\t<tr class="productGrid">\t\r\n\t\t\t<td class="productGrid bld" colspan="1">Oberfläche</td>\r\n\t\t\t<td class="productGrid bld" colspan="1">¾ nitrile coating</td>\r\n\t\t\t</tr>\r\n\t\t<tr class="productGrid">\t\r\n\t\t\t<td class="productGrid bld" colspan="1">Messgerät</td>\r\n\t\t\t<td class="productGrid bld" colspan="1">12</td>\r\n\t\t\t</tr>\r\n\t\t<tr class="productGrid">\t\r\n\t\t\t<td class="productGrid bld" colspan="1">Stulpenart</td>\r\n\t\t\t<td class="productGrid bld" colspan="1">Strickbündchen</td>\r\n\t\t\t</tr>\r\n\t\t<tr class="productGrid">\t\r\n\t\t\t<td class="productGrid bld" colspan="1">EN Standard</td>\r\n\t\t\t<td class="productGrid bld" colspan="1">EN 388:2016 (3111X)</td>\r\n\t\t\t</tr>\r\n\t\t<style type="text/css">\r\n \t.stibospecprodgrid {\r\n \t\t\tdisplay:block;\r\n \t\t\toverflow-x:auto;\r\n \t\t\t}\t\r\n</style>\r\n\r\n<script type="text/javascript">\r\n\r\n\t$(document).ready(function() {\r\n\t\tif (numCol("#stiboSpecGridTable") > 4){\r\n\t\t\t$('#stiboSpecGrid').addClass('stibospecprodgrid');\r\n\t\t}\t\t\r\n\t});\r\n\t\r\n\tfunction numCol(table) {\r\n\t var maxColNum = 0;\r\n\t var i=0;\r\n\t var trs = $(table).find("tr");\r\n\t for ( i=0; i<trs.length; i++ ) {\r\n\t maxColNum = Math.max(maxColNum, getColForTr(trs[i]));\r\n\t }\r\n\r\n\t return maxColNum;\r\n\t}\r\n\tfunction getColForTr(tr) {\r\n\r\n\t var tds = $(tr).find("td");\r\n\r\n\t var numCols = 0;\r\n\r\n\t var i=1;\r\n\t for ( i=1; i<tds.length; i++ ) {\r\n\t var span = $(tds[i]).attr("colspan");\r\n\r\n\t if ( span )\r\n\t numCols += parseInt(span);\r\n\t else {\r\n\t numCols++;\r\n\t }\r\n\t }\r\n\t return numCols;\r\n\t}\r\n</script>\t\t\t\t\t\t\t\t\r\n</tbody>\r\n\t\t\t\t\t</table>\r\n\t\t\t\t\t</div>"}]