Drucken
Lamb serum
Lieferant: Biowest
Ratings: (keine Bewertung)
Gesamtbewertung: 0
Bewertung: 0.0 von 5
BWSTS2300-100EA 23 EUR
BWSTS2300-100 BWSTS2300-500 BWSTS230H-500
Lamb serum
Lamb serum is the liquid fraction of whole blood that is collected after the blood is allowed to clot; it is the blood plasma with the fibrinogens removed. Serum includes all proteins not used in blood clotting (coagulation) and all the electrolytes, antibodies, antigens, hormones, and any exogenous substances. Lamb serum is used as a blocking agent in immunoassays and controls.